siRNA Details
Gene Symbol: 9830107B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177896.1 | NP_001171367.1 | 4 | 2838 | ||
NM_001177897.1 | NP_001171368.1 | 4 | 2691 | |||
NM_177824.4 | NP_808492.1 | 3 | 2747 | |||
GenBank mRNA | AK079398.1 | - | 3 | 2708 | ||
EF646801.1 | - | 2539 |
