siRNA Details
Gene Symbol: Thada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183021.3 | NP_898842.2 | 12 | 2039 | ||
GenBank mRNA | AK044550.1 | - | 11 | 1300 | ||
EF222207.1 | - | 12 | 1947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
