siRNA Details
Gene Symbol: Spink10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177829.3 | NP_808497.1 | 8, 9 | 688 | ||
GenBank mRNA | AK039062.1 | - | 8, 9 | 689 | ||
AK162840.1 | - | 8, 9 | 652 | |||
AV380905.2 | - | 3, 4 | 224 | |||
BC049733.1 | - | 345 | ||||
BC125360.1 | - | 8, 9 | 604 | 40129875 | ||
BC125362.1 | - | 8, 9 | 604 | 40129877 | ||
DQ437330.1 | - | 8, 9 | 688 |
