siRNA Details
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028639.1 | - | 4 | 922 | ||
BC141310.1 | - | 4 | 760 | 9055922 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028639.1 | - | 4 | 922 | ||
BC141310.1 | - | 4 | 760 | 9055922 |