siRNA Details
Gene Symbol: E130311K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177856.4 | NP_808524.1 | 5 | 663 | ||
GenBank mRNA | AK087526.1 | - | 5 | 664 | ||
BC125500.1 | - | 5 | 750 | 40130015 | ||
BC132160.1 | - | 5 | 750 | 40130437 |
