siRNA Details
Gene Symbol: Skint9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177864.2 | NP_808532.1 | 7 | 1145 | ||
GenBank mRNA | AK037244.1 | - | 7 | 1145 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Skint9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177864.2 | NP_808532.1 | 7 | 1145 | ||
GenBank mRNA | AK037244.1 | - | 7 | 1145 |