siRNA Details
Gene Symbol: Fhad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177868.4 | NP_808536.2 | 2 | 202 | ||
GenBank mRNA | AK045799.1 | - | 4 | 532 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003750075, XM_006239329, XM_006239330.
Gene Ontology Categories:
