siRNA Details
Gene Symbol: Fhad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177868.4 | NP_808536.2 | 5, 6 | 903 | ||
GenBank mRNA | AK045799.1 | - | 7, 8 | 1233 |

More Information
Other Species Targeted

Gene Ontology Categories:
