siRNA Details
Gene Symbol: 4933411G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177880.4 | NP_808548.1 | 7 | 1184 | ||
GenBank mRNA | AK030175.1 | - | 7 | 1185 | ||
BC115568.1 | - | 7 | 717 | 40102090 | ||
BC115569.1 | - | 7 | 806 | 40102091 |
