siRNA Details
Gene Symbol: Gm16938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085722.1 | - | 987 | |||
BC030866.1 | - | 1 | 223 | |||
BC040816.1 | - | 1 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
