siRNA Details
Gene Symbol: Wdr72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033500.3 | NP_001028672.2 | 2 | 485 | ||
GenBank mRNA | AK052055.1 | - | 2 | 489 | ||
AK135824.1 | - | 2 | 465 | |||
AK142559.1 | - | 2 | 465 | |||
BB658361.1 | - | 2 | 485 | |||
BC145956.1 | - | 2 | 371 | |||
BC145958.1 | - | 2 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
