siRNA Details
Gene Symbol: GLYAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005838.3 | NP_005829.3 | 2, 3 | 244 | ||
NM_201648.2 | NP_964011.2 | 2, 3 | 244 | |||
GenBank mRNA | AB013093.1 | - | 2, 3 | 193 | ||
AB665285.1 | - | 2, 3 | 222 | |||
AF023466.1 | - | 2, 3 | 173 | |||
BC009785.1 | - | 2, 3 | 248 | 4045672 | ||
BG401676.1 | - | 2, 3 | 284 | |||
BG617408.1 | - | 2, 3 | 226 | |||
BP277146.1 | - | 2, 3 | 246 | |||
CB161978.1 | - | 2, 3 | 168 | |||
CR456927.1 | - | 1, 2 | 86 | |||
DR004166.1 | - | 2, 3 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
