siRNA Details
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172887.2 | NP_766475.2 | 55 | 8415 | ||
GenBank mRNA | AK054402.1 | - | 5 | 841 | ||
AK079097.1 | - | 7 | 899 | |||
AK220506.1 | - | 13 | 2046 | |||
BC042756.1 | - | 7 | 984 | |||
BC096532.1 | - | 12 | 1561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
