siRNA Details
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172887.2 | NP_766475.2 | 61 | 10481 | ||
GenBank mRNA | AK041177.1 | - | 1 | 1625 | ||
AK079097.1 | - | 14 | 2981 | |||
AK086572.1 | - | 1 | 1169 | |||
AK220506.1 | - | 20 | 4128 | |||
BC042756.1 | - | 13 | 3053 | |||
BC096532.1 | - | 19 | 3645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
