siRNA Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036792.1 | - | 3 | 678 | ||
AK087075.1 | - | 3 | 683 | |||
AK087630.1 | - | 3 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
