siRNA Details
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178785.3 | NP_848900.2 | 7 | 1070 | ||
GenBank mRNA | AK030797.1 | - | 7 | 1073 | ||
AK040573.1 | - | 7 | 1058 | |||
AK041479.1 | - | 8 | 928 | |||
AK088449.1 | - | 7 | 1032 | |||
AK088987.1 | - | 7 | 1048 | |||
AK131128.1 | - | 9 | 977 | |||
BC132341.1 | - | 9 | 976 | 40130618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
