siRNA Details
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178785.3 | NP_848900.2 | 11 | 1986 | ||
GenBank mRNA | AK030797.1 | - | 11 | 1989 | ||
AK040573.1 | - | 11 | 1974 | |||
AK041479.1 | - | 12 | 1807 | |||
AK088449.1 | - | 11 | 1948 | |||
AK088987.1 | - | 11 | 1964 | |||
AK131128.1 | - | 13 | 1893 | |||
BC132341.1 | - | 13 | 1892 | 40130618 |

More Information
Other Species Targeted

Gene Ontology Categories:
