siRNA Details
Gene Symbol: Acad12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178799.3 | NP_848914.2 | 11 | 2338 | ||
GenBank mRNA | AK033963.1 | - | 11 | 2257 | ||
BB352084.1 | - | 1 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
