siRNA Details
Gene Symbol: 2610524H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181075.3 | NP_851420.1 | 2 | 244 | ||
GenBank mRNA | AK012147.1 | - | 2 | 233 | ||
AK148078.1 | - | 2 | 219 | |||
BC089033.1 | - | 2 | 224 | |||
BC096574.1 | - | 2 | 235 | 1495259 | ||
BY090347.1 | - | 2 | 244 |
Gene Symbol: AW743365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW743365.1 | - | 1 | 105 |
