siRNA Details
Gene Symbol: Aldoart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199270.1 | NP_001186199.1 | 1 | 1339 | ||
GenBank mRNA | AK014956.1 | - | 1 | 1340 | ||
AK016845.1 | - | 1 | 1312 | |||
EF662061.1 | - | 1 | 1275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
