siRNA Details
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181348.4 | NP_851993.3 | 11 | 8487 | ||
GenBank mRNA | AK038997.1 | - | 3 | 251 | ||
AK054190.1 | - | 3 | 250 | |||
AK163576.1 | - | 3 | 240 | |||
AK165582.1 | - | 3 | 303 | |||
AK170001.1 | - | 3 | 181 | |||
AK172939.1 | - | 3 | 240 | |||
BC094224.1 | - | 4 | 2731 | |||
BC095978.1 | - | 3 | 225 | 30726642 | ||
FR693373.1 | - | 3 | 249 | |||
GQ403051.1 | - | 3 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
