siRNA Details
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181348.4 | NP_851993.3 | 11 | 8545 | ||
GenBank mRNA | AK038997.1 | - | 3 | 309 | ||
AK054190.1 | - | 3 | 308 | |||
AK163576.1 | - | 3 | 298 | |||
AK165582.1 | - | 3 | 361 | |||
AK170001.1 | - | 3 | 239 | |||
AK172939.1 | - | 3 | 298 | |||
BC094224.1 | - | 4 | 2789 | |||
BC095978.1 | - | 3 | 283 | 30726642 | ||
FR693373.1 | - | 3 | 307 | |||
GQ403051.1 | - | 3 | 300 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006231152.
Gene Ontology Categories:
