siRNA Details
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181348.4 | NP_851993.3 | 20 | 9632 | ||
GenBank mRNA | AK038997.1 | - | 11 | 1360 | ||
AK054190.1 | - | 9 | 1272 | |||
AK165582.1 | - | 9 | 1325 | |||
AK170001.1 | - | 9 | 1203 | |||
AK172939.1 | - | 12 | 1358 | |||
BC094224.1 | - | 13 | 3876 | |||
BC095978.1 | - | 9 | 1247 | 30726642 | ||
FR693373.1 | - | 11 | 1358 | |||
GQ403051.1 | - | 11 | 1351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
