siRNA Details
Gene Symbol: 4930529C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015930.1 | - | 1 | 282 |
Gene Symbol: Zbed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181412.3 | NP_852077.1 | 3 | 1784 | ||
GenBank mRNA | AK122335.1 | - | 3 | 1754 | ||
BC052174.1 | - | 2 | 1687 | 6518433 | ||
BC092524.1 | - | 1 | 1408 |
