siRNA Details
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050814.1 | - | 2 | 458 | ||
AK139449.1 | - | 2 | 670 | |||
BC052359.1 | - | 2 | 663 | 6585408 | ||
BQ444292.1 | - | 2 | 692 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050814.1 | - | 2 | 458 | ||
AK139449.1 | - | 2 | 670 | |||
BC052359.1 | - | 2 | 663 | 6585408 | ||
BQ444292.1 | - | 2 | 692 |