siRNA Details
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050814.1 | - | 2 | 832 | ||
AK078069.1 | - | 2 | 1089 | |||
AK139449.1 | - | 2 | 1044 | |||
BC052359.1 | - | 2 | 1037 | 6585408 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050814.1 | - | 2 | 832 | ||
AK078069.1 | - | 2 | 1089 | |||
AK139449.1 | - | 2 | 1044 | |||
BC052359.1 | - | 2 | 1037 | 6585408 |