siRNA Details
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050814.1 | - | 2 | 908 | ||
AK139449.1 | - | 2 | 1120 | |||
BC052359.1 | - | 2 | 1113 | 6585408 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050814.1 | - | 2 | 908 | ||
AK139449.1 | - | 2 | 1120 | |||
BC052359.1 | - | 2 | 1113 | 6585408 |