siRNA Details
Gene Symbol: Zfp516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177464.1 | NP_001170935.1 | 6 | 5088 | ||
NM_183033.2 | NP_898854.1 | 6 | 5110 | |||
GenBank mRNA | AK034874.1 | - | 1 | 192 | ||
AK084274.1 | - | 4 | 2064 | |||
AK129092.1 | - | 4 | 2640 | |||
BC023380.1 | - | 1 | 530 | |||
BC053104.1 | - | 5 | 4902 | 6406544 |
