siRNA Details
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183138.2 | NP_898961.2 | 9 | 3092 | ||
GenBank mRNA | AK084164.1 | - | 4 | 445 | ||
BC096437.1 | - | 5 | 606 | |||
HQ423151.1 | - | 9 | 3266 | |||
HQ423152.1 | - | 8 | 3209 | |||
JX946278.1 | - | 10 | 3798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
