siRNA Details
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183138.2 | NP_898961.2 | 10 | 3143 | ||
GenBank mRNA | AK084164.1 | - | 5 | 496 | ||
BC096437.1 | - | 6 | 657 | |||
HQ423151.1 | - | 10 | 3317 | |||
HQ423152.1 | - | 9 | 3260 | |||
JX946278.1 | - | 11 | 3849 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_001057850, XM_008763095, XM_006236794, XM_006236792, XM_006236793, and others.
Gene Ontology Categories:
