siRNA Details
Gene Symbol: BC048679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193274.1 | NP_001180203.1 | 4 | 215 | ||
NM_183143.3 | NP_898966.1 | 4 | 215 | |||
GenBank mRNA | AY226992.1 | - | 4 | 166 | ||
BC048679.1 | - | 4 | 166 | |||
BC145512.1 | - | 4 | 156 | |||
BC145514.1 | - | 3 | 138 | |||
BG803866.1 | - | 3 | 197 |
