siRNA Details
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183172.2 | NP_898995.1 | 9 | 1631 | ||
GenBank mRNA | AY940666.1 | - | 9 | 1600 | ||
BC051080.1 | - | 9 | 1628 | 6490136 |

More Information
Other Species Targeted

Gene Ontology Categories:
