siRNA Details
Gene Symbol: Fat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183221.3 | NP_899044.3 | 17 | 14674 | ||
GenBank mRNA | AK031348.1 | - | 1 | 1240 | ||
BC079887.1 | - | 9 | 5768 | |||
DQ286572.1 | - | 17 | 14674 |

More Information
Other Species Targeted

Gene Ontology Categories:
