siRNA Details
Gene Symbol: Fcrl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113238.1 | NP_001106709.1 | 3 | 313 | ||
NM_183222.3 | NP_899045.3 | 4 | 577 | |||
GenBank mRNA | AK089756.1 | - | 3 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
