siRNA Details
Gene Symbol: Usp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183225.2 | NP_899048.2 | 7 | 1144 | ||
GenBank mRNA | AK045043.1 | - | 7 | 1137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
