siRNA Details
Gene Symbol: Sema3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011348.2 | NP_035478.2 | 17 | 5743 | ||
GenBank mRNA | AK032603.1 | - | 1 | 1215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
