siRNA Details
Gene Symbol: LINC01587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126517.1 | 2, 3 | 488 | |||
NR_126518.1 | 2, 3 | 467 | ||||
NR_126519.1 | 3, 4 | 376 | ||||
GenBank mRNA | BC117444.1 | - | 2, 3 | 421 | 40125995 | |
BC117446.1 | - | 2, 3 | 421 | 40125997 | ||
BX094519.1 | - | 2, 3 | 285 | |||
D82070.1 | - | 2, 3 | 467 | |||
DB043858.1 | - | 3, 4 | 376 |
