siRNA Details
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198632.2 | NP_941034.2 | 1 | 1547 | ||
GenBank mRNA | AK051455.1 | - | 1 | 1549 | ||
AK220282.1 | - | 1 | 994 | |||
BC094596.1 | - | 1 | 1474 | 5705515 |

More Information
Other Species Targeted

Gene Ontology Categories:
