siRNA Details
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198632.2 | NP_941034.2 | 1 | 1616 | ||
GenBank mRNA | AK051455.1 | - | 1 | 1618 | ||
AK220282.1 | - | 1 | 1063 | |||
BC094596.1 | - | 1 | 1543 | 5705515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
