siRNA Details
Gene Symbol: Zfp335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199027.2 | NP_950192.2 | 10 | 1903 | ||
GenBank mRNA | AK157577.1 | - | 10 | 1734 | ||
BC062921.1 | - | 10 | 1903 | |||
BC151013.1 | - | 9 | 1643 | 9087926 | ||
BC157954.1 | - | 9 | 1643 | 9088127 | ||
BC172135.1 | - | 9 | 1643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
