siRNA Details
Gene Symbol: Zfp335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199027.2 | NP_950192.2 | 24, 25 | 3946 | ||
GenBank mRNA | AK157577.1 | - | 24, 25 | 3777 | ||
AK211288.1 | - | 1, 2 | 73 | |||
BC062921.1 | - | 23, 24 | 3869 | |||
BC151013.1 | - | 23, 24 | 3686 | 9087926 | ||
BC172135.1 | - | 22, 23 | 3765 |

More Information
Other Species Targeted

Gene Ontology Categories:
