siRNA Details
Gene Symbol: Tas2r135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199159.1 | NP_954610.1 | 1 | 841 | ||
GenBank mRNA | BC115532.1 | - | 1 | 837 | 40101425 | |
BC115533.1 | - | 1 | 837 | 40101426 | ||
BC139470.1 | - | 1 | 936 | 8862492 | ||
BC139473.1 | - | 1 | 936 | 8862495 |
