siRNA Details
Gene Symbol: Cep162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199316.2 | NP_955020.2 | 7 | 922 | ||
GenBank mRNA | AK129260.1 | - | 3 | 255 | ||
AK135482.1 | - | 7 | 922 | |||
AK163175.1 | - | 7 | 909 | |||
BC004073.1 | - | 7 | 681 | |||
BC064713.1 | - | 7 | 678 | |||
BC150998.1 | - | 7 | 727 | 9087911 | ||
BC151006.1 | - | 7 | 727 | 9087919 | ||
BC151104.1 | - | 7 | 727 | 9088017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
