siRNA Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024457.2 | NP_077777.1 | 7, 8 | 757 | ||
GenBank mRNA | AK143910.1 | - | 7, 8 | 740 | ||
AK153870.1 | - | 7, 8 | 714 | |||
AK163242.1 | - | 7, 8 | 821 | |||
AK167548.1 | - | 7, 8 | 713 | |||
BC033382.1 | - | 7, 8 | 697 | |||
BC052480.1 | - | 7, 8 | 757 | |||
BC094066.1 | - | 7, 8 | 697 | |||
BC147390.1 | - | 7, 8 | 629 | 9056464 | ||
BC147391.1 | - | 7, 8 | 629 | 9056441 | ||
BC147700.1 | - | 6, 7 | 615 | 9087459 | ||
BC147713.1 | - | 6, 7 | 615 | 9087472 | ||
CK793274.1 | - | 2, 3 | 274 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008765377, FQ229455, U07795, BC081731.
Gene Ontology Categories:
