siRNA Details
Gene Symbol: Prdm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144809.2 | NP_659058.2 | 7 | 832 | ||
GenBank mRNA | AB844106.1 | - | 7 | 832 | ||
AB844107.1 | - | 7 | 832 | |||
AB844108.1 | - | 7 | 832 | |||
AB844109.1 | - | 7 | 838 | |||
AB844110.1 | - | 7 | 832 | |||
AB844111.1 | - | 7 | 810 | |||
AB844112.1 | - | 7 | 810 | |||
AB844113.1 | - | 7 | 832 | |||
AB844114.1 | - | 7 | 832 | |||
AB844116.1 | - | 835 | ||||
BC012016.1 | - | 9 | 973 | |||
BC023014.1 | - | 2 | 494 | 5365459 | ||
BC049903.1 | - | 9 | 973 | 4501059 | ||
FJ212287.1 | - | 7 | 810 | |||
GU216230.1 | - | 7 | 810 | |||
HQ704390.1 | - | 7 | 810 | |||
HQ704391.1 | - | 7 | 810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
