siRNA Details
Gene Symbol: Rcor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290279.1 | NP_001277208.1 | 10 | 1501 | ||
NM_001290280.1 | NP_001277209.1 | 11 | 1559 | |||
GenBank mRNA | AK040839.1 | - | 10 | 1488 | ||
BC020065.1 | - | 11 | 1445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
