siRNA Details
Gene Symbol: Dnah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033668.1 | NP_001028840.1 | 9 | 1396 | ||
GenBank mRNA | AK053204.1 | - | 9 | 1392 | ||
AK076699.1 | - | 5 | 721 | |||
BC023155.1 | - | 9 | 1422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
