siRNA Details
Gene Symbol: PRDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005809.5 | NP_005800.3 | 6 | 1007 | ||
GenBank mRNA | AK293957.1 | - | 6 | 866 | ||
BC000452.2 | - | 6 | 886 | 2821457 | ||
BC003022.1 | - | 6 | 886 | 2821457 | ||
BC039428.1 | - | 6 | 946 | 5303023 | ||
BC064138.1 | - | 4 | 1689 | |||
EU668325.1 | - | 6 | 886 | |||
X82321.1 | - | 4 | 726 | |||
Z22548.1 | - | 6 | 914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
