siRNA Details
Gene Symbol: Tmem184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161548.1 | NP_001155020.1 | 3, 4 | 633 | ||
NM_144914.3 | NP_659163.3 | 2, 3 | 453 | |||
GenBank mRNA | AK050489.1 | - | 3, 4 | 603 | ||
AK148454.1 | - | 425 | ||||
BC026659.1 | - | 3, 4 | 592 | 4988857 |

More Information
Other Species Targeted

Gene Ontology Categories:
