siRNA Details
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285988.1 | NP_001272917.1 | 2, 3 | 327 | ||
NM_001285989.1 | NP_001272918.1 | 2, 3 | 327 | |||
NM_001285990.1 | NP_001272919.1 | 2, 3 | 327 | |||
NM_145436.2 | NP_663411.2 | 2, 3 | 327 | |||
GenBank mRNA | AK039377.1 | - | 2, 3 | 124 | ||
AK134425.1 | - | 1, 2 | 39 | |||
AK148278.1 | - | 2, 3 | 183 | |||
AK161471.1 | - | 2, 3 | 196 | |||
BC023187.1 | - | 2, 3 | 185 | |||
BC157955.1 | - | 2, 3 | 326 | 9088128 | ||
BC172100.1 | - | 2, 3 | 326 | 9054386 | ||
BC172149.1 | - | 2, 3 | 326 | |||
BY036319.1 | - | 2, 3 | 328 |

More Information
Other Species Targeted

Gene Ontology Categories:
